Gene Transcripts

Gene Symbol
LOC440896
Entrez Gene ID
440896
Gene Name
uncharacterized LOC440896
Gene Aliases
Chromosome Location
Chr. 9: 41100985 - 41107812 on Build GRCh38
UniGene ID
Hs.643596
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015361.1 2 236
GenBank mRNA AK055091.1 - 2 239
AK127288.1 - 2 1352
BC032955.1 - 5 895


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB043593.1 - 2 220 Chr. 9 - 41105012 - 41107791 [-]
GenBankmRNA DB057962.1 - 2 215 Chr. 9 - 41105045 - 41107791 [-]
GenBankmRNA DB088025.1 - 2 275 Chr. 9 - 41105091 - 41107851 [-]
GenBankmRNA DB041032.1 - 2 223 Chr. 9 - 41105030 - 41107791 [-]
GenBankmRNA DA196424.1 - 2 220 Chr. 9 - 41105011 - 41107796 [-]
GenBankmRNA DA198144.1 - 2 239 Chr. 9 - 41105098 - 41107812 [-]
GenBankmRNA DB091999.1 - 2 214 Chr. 9 - 41106033 - 41107789 [-]
GenBankmRNA DB464362.1 - 2 234 Chr. 9 - 41106178 - 41107810 [-]
GenBankmRNA DB067669.1 - 2 215 Chr. 9 - 41106019 - 41107791 [-]
GenBankmRNA DB474974.1 - 2 234 Chr. 9 - 41105100 - 41107810 [-]
GenBankmRNA DB475054.1 - 2 234 Chr. 9 - 41105106 - 41107810 [-]
GenBankmRNA DB442482.1 - 2 271 Chr. 9 - 41106209 - 41107847 [-]

View Full Product Details