Gene Transcripts
- Gene Symbol
- LOC101928509
- Entrez Gene ID
- 101928509
- Gene Name
- uncharacterized LOC101928509
- Gene Aliases
- Chromosome Location
- Chr. 4: 174094660 - 174220398 on Build GRCh38
- UniGene ID
- Hs.147719
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125896.1 |
|
5 |
591 |
|
GenBank mRNA |
AK125257.1 |
- |
5 |
591 |
|
View Full Product Details