Gene Transcripts
- Gene Symbol
- DKFZP434L187
- Entrez Gene ID
- 26082
- Gene Name
- uncharacterized LOC26082
- Gene Aliases
- Chromosome Location
- Chr. 15: 30196036 - 30214540 on Build GRCh38
- UniGene ID
- Hs.652128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026771.1 |
|
3 |
3893 |
|
GenBank mRNA |
AL117445.1 |
- |
2 |
3782 |
|
View Full Product Details