Gene Transcripts
- Gene Symbol
- LINC00592
- Entrez Gene ID
- 283404
- Gene Name
- long intergenic non-protein coding RNA 592
- Gene Aliases
- Chromosome Location
- Chr. 12: 52210930 - 52223813 on Build GRCh38
- UniGene ID
- Hs.662443
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027358.1 |
|
3 |
302 |
|
GenBank mRNA |
BC039104.1 |
- |
3 |
302 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG721705.1 |
- |
3 |
307 |
|
Chr. 12 - 52210928 - 52212902 [+] |
View Full Product Details