Gene Transcripts
- Gene Symbol
- LOC101929153
- Entrez Gene ID
- 101929153
- Gene Name
- uncharacterized LOC101929153
- Gene Aliases
- Chromosome Location
- Chr. 5: 4773481 - 4774865 on Build GRCh38
- UniGene ID
- Hs.385526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104619.1 |
|
2 |
240 |
|
GenBank mRNA |
BC034630.1 |
- |
2 |
211 |
|
BU852927.1 |
- |
2 |
241 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB457696.1 |
- |
2 |
189 |
|
Chr. 5 - 4773532 - 4774542 [+] |
GenBankmRNA |
BG772454.1 |
- |
2 |
216 |
|
Chr. 5 - 4773508 - 4774853 [+] |
GenBankmRNA |
DB523186.1 |
- |
2 |
189 |
|
Chr. 5 - 4773532 - 4774539 [+] |
View Full Product Details