Gene Transcripts

Gene Symbol
ROPN1L
Entrez Gene ID
83853
Gene Name
rhophilin associated tail protein 1 like
Gene Aliases
ASP, RSPH11
Chromosome Location
Chr. 5: 10441862 - 10482807 on Build GRCh38
UniGene ID
Hs.381089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006714503.2 XP_006714566.1 6 1317
XM_006714504.2 XP_006714567.1 7 1219

View Full Product Details