Gene Transcripts
- Gene Symbol
- LOC101929698
- Entrez Gene ID
- 101929698
- Gene Name
- uncharacterized LOC101929698
- Gene Aliases
- Chromosome Location
- Chr. 20: 32509959 - 32520285 on Build GRCh38
- UniGene ID
- Hs.638392
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110619.1 |
|
1 |
69 |
|
GenBank mRNA |
AK097865.1 |
- |
1 |
69 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR610740.1 |
- |
1 |
21 |
|
Chr. 20 - 32510007 - 32511863 [+] |
View Full Product Details