Gene Transcripts

Gene Symbol
Cts3
Entrez Gene ID
117066
Gene Name
cathepsin 3
Gene Aliases
1600000I23Rik
Chromosome Location
Chr. 13: 61564630 - 61570162 on Build GRCm38
UniGene ID
Mm.485905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026906.3 NP_081182.2 7 1016
NM_054092.2 NP_473433.2 7 941
GenBank mRNA AF456464.1 - 6 801
AK005389.1 - 7 939
AY034574.1 - 7 955
BC099388.1 - 7 910 30786828

View Full Product Details