Gene Transcripts

Gene Symbol
Steap4
Entrez Gene ID
117167
Gene Name
STEAP family member 4
Gene Aliases
1110021O17Rik, AI481214, Tiarp, Tnfaip9
Chromosome Location
Chr. 5: 7960472 - 7982213 on Build GRCm38
UniGene ID
Mm.31403
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054098.3 NP_473439.2 3 809
GenBank mRNA AJ319746.1 - 3 812
AK040760.1 - 3 825
AY029778.1 - 3 788
BC006651.1 - 3 809 3500056

View Full Product Details