Gene Transcripts

Gene Symbol
Slc7a12
Entrez Gene ID
140918
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 12
Gene Aliases
Asc-2, XAT1
Chromosome Location
Chr. 3: 14480691 - 14505819 on Build GRCm38
UniGene ID
Mm.215250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080852.2 NP_543128.1 3 997
GenBank mRNA AB055005.1 - 3 1002

View Full Product Details