Gene Transcripts

Gene Symbol
PCBP2
Entrez Gene ID
5094
Gene Name
poly(rC) binding protein 2
Gene Aliases
HNRNPE2, HNRPE2, hnRNP-E2
Chromosome Location
Chr. 12: 53452102 - 53481162 on Build GRCh38
UniGene ID
Hs.546271
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098620.2 NP_001092090.1 10 1010
NM_001128911.1 NP_001122383.1 11 1115
NM_001128912.1 NP_001122384.1 11 1103
NM_001128913.1 NP_001122385.1 10 1022
NM_001128914.1 NP_001122386.1 10 1010
NM_005016.5 NP_005007.2 11 1115
NM_031989.4 NP_114366.1 11 1103
GenBank mRNA AB188306.1 - 10 1010
AB208825.1 - 10 837
AK023529.1 - 6 1714
AK130583.1 - 4 257
AK225357.1 - 9 724
AK292141.1 - 11 880
AK299210.1 - 7 574
AK302067.1 - 9 739
BC001155.1 - 10 841 3356341
BC035420.1 - 11 1088
BC071942.1 - 11 1105 6424433
BC107688.1 - 9 688 5395926
X78136.1 - 10 787

View Full Product Details