Gene Transcripts
- Gene Symbol
- Vmn1r210
- Entrez Gene ID
- 171269
- Gene Name
- vomeronasal 1 receptor 210
- Gene Aliases
- V1rh10
- Chromosome Location
- Chr. 13: 22827194 - 22828114 on Build GRCm38
- UniGene ID
- Mm.377221
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134235.1 |
NP_598996.1
|
1 |
170 |
|
- Gene Symbol
- Vmn1r205
- Entrez Gene ID
- 171251
- Gene Name
- vomeronasal 1 receptor 205
- Gene Aliases
- V1rh8
- Chromosome Location
- Chr. 13: 22591980 - 22592930 on Build GRCm38
- UniGene ID
- Mm.377206
- Species
- Mus musculus
View Full Product Details