Gene Transcripts
- Gene Symbol
- Vmn1r217
- Entrez Gene ID
- 171273
- Gene Name
- vomeronasal 1 receptor 217
- Gene Aliases
- V1rh14
- Chromosome Location
- Chr. 13: 23113834 - 23114730 on Build GRCm38
- UniGene ID
- Mm.377225
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134239.1 |
NP_599000.1
|
1 |
792 |
|
- Gene Symbol
- Vmn1r220
- Entrez Gene ID
- 171271
- Gene Name
- vomeronasal 1 receptor 220
- Gene Aliases
- V1rh12
- Chromosome Location
- Chr. 13: 23183628 - 23187867 on Build GRCm38
- UniGene ID
- Mm.377223
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134237.1 |
NP_598998.1
|
1 |
792 |
|
View Full Product Details