Gene Transcripts

Gene Symbol
Gan
Entrez Gene ID
209239
Gene Name
giant axonal neuropathy
Gene Aliases
A330045G18, gigaxonin
Chromosome Location
Chr. 8: 117158135 - 117205186 on Build GRCm38
UniGene ID
Mm.132992
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081151.1 NP_001074620.1 9, 10 1513
GenBank mRNA AK039455.1 - 9, 10 1512
BC112432.1 - 9, 10 1512

View Full Product Details