Gene Transcripts

Gene Symbol
Tsga10
Entrez Gene ID
211484
Gene Name
testis specific 10
Gene Aliases
4933432N21Rik, AI843383, Gm217, Mtsga10
Chromosome Location
Chr. 1: 37728144 - 37866739 on Build GRCm38
UniGene ID
Mm.332756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290720.1 NP_001277649.1 7 1070
NM_001290721.1 NP_001277650.1 6 877
NM_207228.3 NP_997111.1 7 937
GenBank mRNA AF530050.3 - 6 882
AK030254.1 - 7 1070
AK163138.1 - 7 884
AK164954.1 - 7 904
AK170053.1 - 7 960
AY618264.1 - 6 882
BC053407.1 - 6 860
BC066782.1 - 7 934 6705127

View Full Product Details