Gene Transcripts
- Gene Symbol
- Hk3
- Entrez Gene ID
- 212032
- Gene Name
- hexokinase 3
- Gene Aliases
- HK III, HK-III
- Chromosome Location
- Chr. 13: 55005985 - 55021472 on Build GRCm38
- UniGene ID
- Mm.267479
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033245.4 |
NP_001028417.1
|
12, 13 |
2003 |
|
NM_001206390.1 |
NP_001193319.1
|
12, 13 |
1813 |
|
NM_001206391.1 |
NP_001193320.1
|
12, 13 |
1678 |
|
NM_001206392.1 |
NP_001193321.1
|
11, 12 |
1883 |
|
GenBank mRNA |
AK140444.1 |
- |
12, 13 |
1867 |
|
AK149651.1 |
- |
12, 13 |
1853 |
|
AK149992.1 |
- |
12, 13 |
1815 |
|
AK152839.1 |
- |
6, 7 |
1107 |
|
AK154603.1 |
- |
12, 13 |
1975 |
|
AK155701.1 |
- |
12, 13 |
1860 |
|
AK159857.1 |
- |
11, 12 |
1729 |
|
AK159906.1 |
- |
12, 13 |
1913 |
|
AK160018.1 |
- |
12, 13 |
1913 |
|
AK160063.1 |
- |
12, 13 |
1913 |
|
AK162637.1 |
- |
12, 13 |
1946 |
|
AK163658.1 |
- |
12, 13 |
1867 |
|
AK170813.1 |
- |
11, 12 |
1733 |
|
AK171214.1 |
- |
12, 13 |
1941 |
|
AK171248.1 |
- |
12, 13 |
1853 |
|
AK171580.1 |
- |
12, 13 |
1953 |
|
BC117860.1 |
- |
12, 13 |
1898 |
40095265 |
BC117861.1 |
- |
12, 13 |
1763 |
40095267 |
View Full Product Details