Gene Transcripts

Gene Symbol
BC027231
Entrez Gene ID
212547
Gene Name
cDNA sequence BC027231
Gene Aliases
Nepro
Chromosome Location
Chr. 16: 44724301 - 44737284 on Build GRCm38
UniGene ID
Mm.310573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145972.4 NP_666084.1 9 1435
GenBank mRNA AK144612.1 - 9 1416
AK162291.1 - 8 1277
BC027231.1 - 9 1438 3585175

View Full Product Details