Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144810.5 | NP_659059.1 | 1 | 891 | |
GenBank mRNA | AK043039.1 | - | 1 | 229 | |
AK044318.1 | - | 1 | 467 | ||
AK080356.1 | - | 1 | 891 | ||
BC011135.1 | - | 1 | 456 | 4191503 | |
BC023173.1 | - | 1 | 90 |