Gene Transcripts

Gene Symbol
Synrg
Entrez Gene ID
217030
Gene Name
synergin, gamma
Gene Aliases
Ap1gbp1, SYNG
Chromosome Location
Chr. 11: 83964380 - 84044578 on Build GRCm38
UniGene ID
Mm.441722
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194341.2 NP_919322.1 8 743
GenBank mRNA BC056370.1 - 8 703

View Full Product Details