Gene Transcripts

Gene Symbol
Tmc6
Entrez Gene ID
217353
Gene Name
transmembrane channel-like gene family 6
Gene Aliases
D11Ertd204e, EVER1
Chromosome Location
Chr. 11: 117765985 - 117782193 on Build GRCm38
UniGene ID
Mm.286963
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145439.2 NP_663414.3 8 1004
NM_181321.3 NP_851838.1 8 942
GenBank mRNA AK052814.1 - 8 944
AK077671.1 - 6 749
AK090179.1 - 8 961
AK184505.1 - 2 177
AY236497.1 - 8 903
AY263158.1 - 8 929
BC004840.1 - 8 929 2648849
BC006956.1 - 6 714
BC058195.1 - 8 929 5356348

View Full Product Details