Gene Transcripts

Gene Symbol
Eif4e1b
Entrez Gene ID
218268
Gene Name
eukaryotic translation initiation factor 4E family member 1B
Gene Aliases
Eif4eloo, Gm273
Chromosome Location
Chr. 13: 54783927 - 54788459 on Build GRCm38
UniGene ID
Mm.425821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033269.3 NP_001028441.1 2 278
NM_001039683.2 NP_001034772.1 2 278
NM_001286178.1 NP_001273107.1 2 207
NM_001286179.1 NP_001273108.1 2 210
NM_001286180.1 NP_001273109.1 2 207
GenBank mRNA AK139577.1 - 1 516
BC139143.1 - 2 245 8862165
BC139144.1 - 2 242 8862166
DQ279473.1 - 2 273
DQ279474.1 - 2 256
DQ279475.1 - 1 523
DQ279476.1 - 2 438
DV653822.1 - 2 204
DV659735.1 - 2 207
DV662251.1 - 2 207
DV662389.1 - 2 204

View Full Product Details