Gene Transcripts

Gene Symbol
Wdr41
Entrez Gene ID
218460
Gene Name
WD repeat domain 41
Gene Aliases
B830029I03Rik, MSTP048
Chromosome Location
Chr. 13: 94976213 - 95023316 on Build GRCm38
UniGene ID
Mm.33856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172590.3 NP_766178.2 14 1528
GenBank mRNA AK046853.1 - 14 1497
AK149993.1 - 14 1494
AK183715.1 - 2 159

View Full Product Details