Gene Transcripts

Gene Symbol
Ttc7
Entrez Gene ID
225049
Gene Name
tetratricopeptide repeat domain 7
Gene Aliases
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea
Chromosome Location
Chr. 17: 87282082 - 87381770 on Build GRCm38
UniGene ID
Mm.77396
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028639.3 NP_082915.1 12 1768
GenBank mRNA AK040578.1 - 12 1771
AK040610.1 - 12 1739
AK050263.1 - 12 1755
AK080284.1 - 12 1747
AK153704.1 - 12 1750
AK165244.1 - 5 430
BC023773.1 - 8 726
BC042512.1 - 12 1445 4223801

View Full Product Details