Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321378.1 | NP_001308307.1 | 26 | 4246 | |
NM_001321380.1 | NP_001308309.1 | 24 | 3047 | ||
NM_002645.3 | NP_002636.2 | 25 | 4177 | ||
XM_011520186.2 | XP_011518488.1 | 24 | 4004 | ||
XM_017017924.1 | XP_016873413.1 | 26 | 4193 | ||
GenBank mRNA | AK294416.1 | - | 24 | 3034 | |
BC113658.1 | - | 24 | 4029 | 8322710 | |
BX648778.1 | - | 15 | 2113 | ||
Y13367.1 | - | 24 | 3963 |