Gene Transcripts

Gene Symbol
Snx32
Entrez Gene ID
225861
Gene Name
sorting nexin 32
Gene Aliases
B930037P14Rik, Snx6b
Chromosome Location
Chr. 19: 5495274 - 5510694 on Build GRCm38
UniGene ID
Mm.20071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024560.2 NP_001019731.1 8 844
GenBank mRNA AK037118.1 - 835
AK047234.1 - 8 820
AK143441.1 - 7 956
BC048922.1 - 8 844 6311864

View Full Product Details