Gene Transcripts

Gene Symbol
Ndufs8
Entrez Gene ID
225887
Gene Name
NADH dehydrogenase (ubiquinone) Fe-S protein 8
Gene Aliases
BC021616, CI-23kD, TYKY
Chromosome Location
Chr. 19: 3908863 - 3912774 on Build GRCm38
UniGene ID
Mm.44227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271443.1 NP_001258372.1 6 503
NM_001271444.1 NP_001258373.1 6 743
NM_144870.5 NP_659119.2 6 746
GenBank mRNA AK135069.1 - 6 504
AK149674.1 - 6 704
AK153182.1 - 6 690
AK153900.1 - 6 480
AK164901.1 - 6 480
AK167907.1 - 6 481
BC021616.1 - 6 464 5132866
BC086766.1 - 6 457 6820786

View Full Product Details