Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
226043
Gene Name
COBW domain containing 1
Gene Aliases
AV349248, BC018472
Chromosome Location
Chr. 19: 24919916 - 24961616 on Build GRCm38
UniGene ID
Mm.272127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146097.3 NP_666209.1 5 557
GenBank mRNA AK076027.1 - 5 512
AK132291.1 - 5 543
BC018472.1 - 5 499 3661842
BC053745.1 - 5 592 6505520

View Full Product Details