Gene Transcripts

Gene Symbol
Erlin1
Entrez Gene ID
226144
Gene Name
ER lipid raft associated 1
Gene Aliases
2810439N09Rik, C80197, Keo4, Spfh1
Chromosome Location
Chr. 19: 44034943 - 44070493 on Build GRCm38
UniGene ID
Mm.279865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164359.1 NP_001157831.1 6 546
NM_001164360.1 NP_001157832.1 6 454
NM_145502.3 NP_663477.3 5 650
GenBank mRNA AK133557.1 - 5 393
AK145990.1 - 6 427
BC011220.1 - 6 418 4208238
BC023849.1 - 5 513 5345801

View Full Product Details