Gene Transcripts
- Gene Symbol
- Epb4.1l5
- Entrez Gene ID
- 226352
- Gene Name
- erythrocyte protein band 4.1 like 5
- Gene Aliases
- 1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
- Chromosome Location
- Chr. 1: 119545032 - 119649136 on Build GRCm38
- UniGene ID
- Mm.253156
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113416.1 |
NP_001106887.1
|
3, 4 |
509 |
|
NM_145506.4 |
NP_663481.2
|
3, 4 |
509 |
|
GenBank mRNA |
AK027983.1 |
- |
3, 4 |
463 |
|
AK034676.1 |
- |
3, 4 |
431 |
|
AK040354.1 |
- |
3, 4 |
477 |
|
AK044959.1 |
- |
3, 4 |
447 |
|
AK044996.1 |
- |
3, 4 |
455 |
|
AK047523.1 |
- |
3, 4 |
436 |
|
AK079301.1 |
- |
3, 4 |
460 |
|
AK136125.1 |
- |
3, 4 |
464 |
|
AK161141.1 |
- |
3, 4 |
460 |
|
AK167449.1 |
- |
3, 4 |
509 |
|
AK173198.1 |
- |
3, 4 |
443 |
|
BC003937.1 |
- |
3, 4 |
494 |
3486543 |
BC011476.1 |
- |
3, 4 |
443 |
4014009 |
BY737035.1 |
- |
3, 4 |
509 |
|
View Full Product Details