Gene Transcripts

Gene Symbol
R3hdm1
Entrez Gene ID
226412
Gene Name
R3H domain containing 1
Gene Aliases
R3hdm
Chromosome Location
Chr. 1: 128103293 - 128237738 on Build GRCm38
UniGene ID
Mm.221041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181750.2 NP_861415.2 4, 5 641
GenBank mRNA AK051765.1 - 4, 5 619
BC049181.1 - 4, 5 623
BC137772.1 - 3, 4 386 8860791
BC144899.1 - 3, 4 386

View Full Product Details