Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013795.5 | NP_038823.2 | 3 | 350 | |
GenBank mRNA | AK005175.1 | - | 3 | 297 | |
AK005321.1 | - | 3 | 327 | ||
AK005380.1 | - | 3 | 328 | ||
AK008002.1 | - | 3 | 287 | ||
AK008012.1 | - | 3 | 324 | ||
AK008030.1 | - | 3 | 313 | ||
AK008036.1 | - | 3 | 327 | ||
AK008087.1 | - | 3 | 301 | ||
AK008169.1 | - | 3 | 327 | ||
AK008183.1 | - | 3 | 315 | ||
AK008251.1 | - | 3 | 315 | ||
AK008321.1 | - | 3 | 323 | ||
AK009465.1 | - | 3 | 316 | ||
AK009468.1 | - | 3 | 316 | ||
AK009471.1 | - | 3 | 316 | ||
AK009612.1 | - | 3 | 309 | ||
AK009806.1 | - | 3 | 315 | ||
AK011038.1 | - | 3 | 294 | ||
AK011046.1 | - | 3 | 294 | ||
AK011099.1 | - | 3 | 321 | ||
AK011842.1 | - | 3 | 287 | ||
AK019192.1 | - | 2 | 154 | ||
AK019252.1 | - | 3 | 316 | ||
AK019274.1 | - | 3 | 316 | ||
AK019320.1 | - | 3 | 323 | ||
AK019343.1 | - | 3 | 287 | ||
AK019350.1 | - | 3 | 315 | ||
AK019355.1 | - | 3 | 315 | ||
AK146520.1 | - | 3604 | |||
BC031384.1 | - | 3 | 323 | 5356491 | |
BC086880.1 | - | 3 | 286 | 6762989 | |
BC089555.1 | - | 3 | 326 | 30312783 | |
BC094407.1 | - | 3 | 280 | 573475 | |
BY269662.1 | - | 3 | 351 | ||
Y17223.1 | - | 3 | 280 |