Gene Transcripts

Gene Symbol
Ebf4
Entrez Gene ID
228598
Gene Name
early B cell factor 4
Gene Aliases
Ebf3, O/E-4, Olf-1
Chromosome Location
Chr. 2: 130295249 - 130370481 on Build GRCm38
UniGene ID
Mm.32582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110513.1 NP_001103983.1 3 396
GenBank mRNA AF387630.1 - 3 396
AF387631.1 - 3 396
AF387632.1 - 3 396
AF387633.1 - 3 396
AF387634.1 - 3 396
BC138119.1 - 3 396 8861138
BC145155.1 - 4 482 9053706

View Full Product Details