Gene Transcripts

Gene Symbol
Kiz
Entrez Gene ID
228730
Gene Name
kizuna centrosomal protein
Gene Aliases
Gm114, Ncrna00153, Plk1s1
Chromosome Location
Chr. 2: 146855824 - 146970098 on Build GRCm38
UniGene ID
Mm.292352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033298.3 NP_001028470.1 11 1838
GenBank mRNA AK135486.1 - 11 1841
AK218233.1 - 2 111
BC150844.1 - 11 1883 9087755
BC150845.1 - 11 1883 9087756

View Full Product Details