Gene Transcripts

Gene Symbol
Slc35a3
Entrez Gene ID
229782
Gene Name
solute carrier family 35 (UDP-N-acetylglucosamine (UDP-GlcNAc) transporter), member 3
Gene Aliases
2310050P13Rik
Chromosome Location
Chr. 3: 116669469 - 116713960 on Build GRCm38
UniGene ID
Mm.190758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144902.3 NP_659151.1 8 1149
GenBank mRNA AK029398.1 - 8 1521
AK041195.1 - 9 1269
AK049484.1 - 8 1149
AK161317.1 - 8 1107

View Full Product Details