Gene Transcripts

Gene Symbol
Ndufb6
Entrez Gene ID
230075
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6
Gene Aliases
Gm137
Chromosome Location
Chr. 4: 40270663 - 40279368 on Build GRCm38
UniGene ID
Mm.1103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033305.2 NP_001028477.1 1 171
GenBank mRNA AK146762.1 - 1 173
BC147062.1 - 1 160 9056053
BC147063.1 - 1 160 9056059

View Full Product Details