Gene Transcripts

Gene Symbol
Tardbp
Entrez Gene ID
230908
Gene Name
TAR DNA binding protein
Gene Aliases
1190002A23Rik, C85084, TDP-43, Tdp43
Chromosome Location
Chr. 4: 148612382 - 148626996 on Build GRCm38
UniGene ID
Mm.22453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003898.3 NP_001003898.1 6 1071
NM_001003899.2 NP_001003899.1 6 1071
NM_001008545.2 NP_001008545.1 6 1071
NM_001008546.2 NP_001008546.1 6 1071
NM_145556.4 NP_663531.1 6 1071
GenBank mRNA AK032199.1 - 6 841
AK045495.1 - 6 841
AK076208.1 - 6 839
AK083312.1 - 6 845
AK133207.1 - 6 848
AK146002.1 - 6 841
AK146997.1 - 7 750
AK153803.1 - 6 845
AK194463.1 - 2 147
AK195880.1 - 2 153
AY319513.1 - 5 741
AY319514.1 - 5 741
AY319515.1 - 5 741
AY319516.1 - 5 741
BC012873.1 - 6 829 4016437
BC025544.1 - 6 842 5321007
BC027105.1 - 6 833 5359082
BC027772.1 - 6 831 5346061
BC029728.1 - 6 832 4950263
BC031126.1 - 6 833 4977114
BC033475.1 - 6 829 4953334
GQ428775.1 - 5 741
GQ428776.1 - 5 741
GQ428777.1 - 5 741
GU250735.1 - 5 741

View Full Product Details