Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033310.3 | NP_001028482.2 | 3, 4 | 653 | |
GenBank mRNA | AK156053.1 | - | 3, 4 | 622 | |
BC021612.1 | - | 1, 2 | 204 | ||
BC138972.1 | - | 3, 4 | 631 | 8861994 | |
BC145486.1 | - | 3, 4 | 635 | ||
BC145728.1 | - | 3, 4 | 631 | 40131031 | |
DV661727.1 | - | 2, 3 | 264 |