Gene Transcripts

Gene Symbol
Ccz1
Entrez Gene ID
231874
Gene Name
CCZ1 vacuolar protein trafficking and biogenesis associated
Gene Aliases
AU022870
Chromosome Location
Chr. 5: 143987909 - 144014853 on Build GRCm38
UniGene ID
Mm.320432
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177682.3 NP_808350.1 7 711
GenBank mRNA AK090008.1 - 7 725
AK146704.1 - 7 744
AK150793.1 - 7 748
AK152328.1 - 7 747
AK152576.1 - 7 747
AK153160.1 - 7 748
AK213761.1 - 4 282
BC048077.1 - 7 711 6308253

View Full Product Details