Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
231915
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
E430026A01Rik
Chromosome Location
Chr. 5: 149184560 - 149215434 on Build GRCm38
UniGene ID
Mm.29402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013378.2 NP_001013396.2 4 729
NM_001115149.1 NP_001108621.1 4 729
NM_001115150.1 NP_001108622.1 3 560
NM_001115151.1 NP_001108623.1 3 560
NM_001115153.1 NP_001108625.1 5 906
NM_001286824.1 NP_001273753.1 4 687
NM_001286825.1 NP_001273754.1 4 671
GenBank mRNA AK050969.1 - 4 713
AK088787.1 - 3 426
AK145409.1 - 5 804
AK146820.1 - 4 554
AK161654.1 - 3 407
AK168702.1 - 4 554
AK168981.1 - 4 581
BC028849.1 - 4 623
BC032291.1 - 1 2699
BC086664.1 - 4 571

View Full Product Details