Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
231915
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
E430026A01Rik
Chromosome Location
Chr. 5: 149184560 - 149215434 on Build GRCm38
UniGene ID
Mm.29402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013378.2 NP_001013396.2 4 931
NM_001115149.1 NP_001108621.1 4 931
NM_001115150.1 NP_001108622.1 3 762
NM_001115151.1 NP_001108623.1 3 762
NM_001115153.1 NP_001108625.1 5 1108
NM_001286824.1 NP_001273753.1 4 889
NM_001286825.1 NP_001273754.1 4 873
GenBank mRNA AK050969.1 - 4 915
AK088787.1 - 3 628
AK145409.1 - 5 1006
AK146820.1 - 4 756
AK161654.1 - 3 609
AK168702.1 - 4 756
AK168981.1 - 4 783
BC028849.1 - 4 825
BC032291.1 - 1 2901
BC086664.1 - 4 773

View Full Product Details