Gene Transcripts

Gene Symbol
Mypop
Entrez Gene ID
232934
Gene Name
Myb-related transcription factor, partner of profilin
Gene Aliases
Myodysa, P42pop
Chromosome Location
Chr. 7: 18990058 - 19001766 on Build GRCm38
UniGene ID
Mm.440069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145579.3 NP_663554.1 3 1902
GenBank mRNA AF364868.1 - 4 1611
BC042748.1 - 2 1141
BC048544.1 - 1 52
M82905.1 - 1 162

View Full Product Details