Gene Transcripts

Gene Symbol
Gas2l2
Entrez Gene ID
237891
Gene Name
growth arrest-specific 2 like 2
Gene Aliases
Chromosome Location
Chr. 11: 83421635 - 83429521 on Build GRCm38
UniGene ID
Mm.340523
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013759.2 NP_001013781.1 6 1614
GenBank mRNA BC147480.1 - 6 1614
BC150847.1 - 6 1614 9087758

View Full Product Details