Gene Transcripts

Gene Symbol
Fam69c
Entrez Gene ID
240479
Gene Name
family with sequence similarity 69, member C
Gene Aliases
B230399E16Rik
Chromosome Location
Chr. 18: 84718987 - 84741731 on Build GRCm38
UniGene ID
Mm.472378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173770.4 NP_776131.1 4 1014
GenBank mRNA AK046504.1 - 4 1015
BC119040.1 - 2 602 8733789
BC119042.1 - 2 602 8733791

View Full Product Details