Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008709.1 | NP_001008709.1 | 3 | 374 | |
NM_002708.3 | NP_002699.1 | 3 | 341 | ||
NM_206873.1 | NP_996756.1 | 2 | 209 | ||
GenBank mRNA | AK098311.1 | - | 5 | 1150 | |
AK127616.1 | - | 4 | 558 | ||
AK313586.1 | - | 3 | 250 | ||
AL557336.3 | - | 3 | 365 | ||
BC001888.1 | - | 3 | 262 | 3546588 | |
BC004482.2 | - | 3 | 282 | 3689198 | |
BC008010.1 | - | 3 | 282 | 3528742 | |
BE278068.1 | - | 2 | 129 | ||
BG480286.1 | - | 3 | 339 | ||
BM799599.1 | - | 2 | 148 | ||
BT006629.1 | - | 3 | 218 | ||
J04759.1 | - | 2 | 152 | ||
M63960.1 | - | 3 | 247 | ||
S57501.1 | - | 3 | 262 | ||
X70848.1 | - | 3 | 245 |