Gene Transcripts

Gene Symbol
Klhdc7a
Entrez Gene ID
242721
Gene Name
kelch domain containing 7A
Gene Aliases
B230308G19Rik
Chromosome Location
Chr. 4: 139962173 - 139968026 on Build GRCm38
UniGene ID
Mm.60400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173427.2 NP_775603.2 1 878
GenBank mRNA AK045732.1 - 1 881
AK050048.1 - 1 465
AK052747.1 - 1 448

View Full Product Details