Gene Transcripts

Gene Symbol
Lrrc38
Entrez Gene ID
242735
Gene Name
leucine rich repeat containing 38
Gene Aliases
A230053A07Rik
Chromosome Location
Chr. 4: 143346981 - 143371028 on Build GRCm38
UniGene ID
Mm.94020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162983.1 NP_001156455.1 1 174
GenBank mRNA AK038658.1 - 1 136
AK078190.1 - 1 148
BC129963.1 - 1 167
BC129964.1 - 1 174

View Full Product Details