Gene Transcripts

Gene Symbol
PPP2R5E
Entrez Gene ID
5529
Gene Name
protein phosphatase 2 regulatory subunit B'epsilon
Gene Aliases
Chromosome Location
Chr. 14: 63373892 - 63543361 on Build GRCh38
UniGene ID
Hs.334868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282179.1 NP_001269108.1 6 1150
NM_001282180.1 NP_001269109.1 6 1153
NM_001282181.1 NP_001269110.1 5 520
NM_001282182.1 NP_001269111.1 5 475
NM_006246.3 NP_006237.1 6 1153
NR_104104.1 5 956
GenBank mRNA AB209811.1 - 6 1106
AB451233.1 - 5 563
AB451358.1 - 5 563
AK299520.1 - 6 1090
AK301796.1 - 5 520
AK313500.1 - 6 1006
AK316430.1 - 5 474
BC013154.1 - 5 949
BC034994.1 - 7 1147
BC048305.1 - 6 886
BC064358.1 - 6 1133
BC073873.1 - 6 1105
BC092477.1 - 6 1123 5267163
BC093766.1 - 6 621 7939611
BC101479.1 - 6 621 8068985
BC143231.1 - 6 618 9051732
BC143234.1 - 6 621 9051735
L76703.1 - 6 1130
Z69029.1 - 6 1068

View Full Product Details