Gene Transcripts

Gene Symbol
Slitrk4
Entrez Gene ID
245446
Gene Name
SLIT and NTRK-like family, member 4
Gene Aliases
6430513G13, D930039I09Rik
Chromosome Location
Chr. X: 64264883 - 64277367 on Build GRCm38
UniGene ID
Mm.101707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178740.4 NP_848855.2 3 756
GenBank mRNA AK032263.1 - 2 646
AK039763.1 - 3 697
AK046301.1 - 3 626
AK053065.1 - 3 757
BC117890.1 - 3 621 40096585
BC117891.1 - 2 554 40096591

View Full Product Details