Gene Transcripts

Gene Symbol
Mllt6
Entrez Gene ID
246198
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 6
Gene Aliases
AI315037, Af17
Chromosome Location
Chr. 11: 97663412 - 97685461 on Build GRCm38
UniGene ID
Mm.23685
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139311.2 NP_647472.2 7, 8 933
GenBank mRNA AK143477.1 - 3, 4 376
AY050217.1 - 7, 8 1080
BY731745.1 - 3, 4 372

View Full Product Details