Gene Transcripts

Gene Symbol
Vmn1r85
Entrez Gene ID
252909
Gene Name
vomeronasal 1 receptor 85
Gene Aliases
V1rj3
Chromosome Location
Chr. 7: 13084289 - 13085215 on Build GRCm38
UniGene ID
Mm.377268
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145847.1 NP_665846.1 1 384
GenBank mRNA BC110463.1 - 1 384
BC110464.1 - 1 384 40040731

View Full Product Details